ENAH2C557341 Enamine

ethyl[2-(2-methoxyethoxy)ethyl]amine

  • MDL number MFCD16107092
  • Cas Number : 188575-78-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009575

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009575
Availability details for
Quantity
  • Availability
  • Add to Cart