ENAH2C557343 Enamine

(4-methoxy-6-methylpyridin-3-yl)methanol

  • MDL number MFCD24073746
  • Cas Number : 915788-15-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009577

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009577
Availability details for
Quantity
  • Availability
  • Add to Cart