ENAH2C557348 Enamine

[(1E)-4-bromobut-1-en-1-yl]benzene

  • MDL number MFCD11936563
  • Cas Number : 7515-41-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009586

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009586
Availability details for
Quantity
  • Availability
  • Add to Cart