ENAH2C55734A Enamine

2-methyl-5-(propan-2-yl)-1H-indole

  • MDL number MFCD21879819
  • Cas Number : 1368912-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009601

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009601
Availability details for
Quantity
  • Availability
  • Add to Cart