ENAH2C557352 Enamine

4a-methyl-decahydroquinoline

  • MDL number MFCD20547679
  • Cas Number : 37442-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009660

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009660
Availability details for
Quantity
  • Availability
  • Add to Cart