ENAH2C557359 Enamine

pyrazolo[1,5-a]pyrazin-4-amine

  • MDL number MFCD31691106
  • Cas Number : 1565419-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009669

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009669
Availability details for
Quantity
  • Availability
  • Add to Cart