ENAH2C55735D Enamine

(2S)-2-amino-4,4-difluorobutanoic acid hydrochloride

  • MDL number MFCD31691108

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164-165
Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009673

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009673
Availability details for
Quantity
  • Availability
  • Add to Cart