ENAH2C55735F Enamine

[3-(propan-2-yl)cyclobutyl]methanol

  • MDL number MFCD18801139
  • Cas Number : 1269291-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009676

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009676
Availability details for
Quantity
  • Availability
  • Add to Cart