ENAH2C557360 Enamine

ethyl 1H,4H,5H,6H,7H-pyrazolo[4,3-c]pyridine-3-carboxylate dihydrochloride

  • MDL number MFCD31667940
  • Cas Number : 2230808-76-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009679

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009679
Availability details for
Quantity
  • Availability
  • Add to Cart