ENAH2C557361 Enamine

(2E)-3-(4-fluorophenyl)-2-methylprop-2-enoic acid

  • MDL number MFCD06207018
  • Cas Number : 210110-49-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151-153
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009689

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009689
Availability details for
Quantity
  • Availability
  • Add to Cart