ENAH2C55736E Enamine

4-(6-oxopiperidin-2-yl)butanoic acid

  • MDL number MFCD20640623
  • Cas Number : 90942-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1721846

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721846
Availability details for
Quantity
  • Availability
  • Add to Cart