ENAH2C557382 Enamine

4-(5-oxo-4,5-dihydro-1H-pyrazol-1-yl)benzoic acid

  • MDL number MFCD20227721
  • Cas Number : 916169-50-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009770

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009770
Availability details for
Quantity
  • Availability
  • Add to Cart