ENAH2C557383 Enamine

methyl 2-(chloromethyl)-4H,5H,6H-pyrrolo[1,2-b]pyrazole-3-carboxylate

  • MDL number MFCD26130366
  • Cas Number : 1707562-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1721894

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721894
Availability details for
Quantity
  • Availability
  • Add to Cart