ENAH2C55738A Enamine

methyl 2-(chloromethyl)-4H,5H,6H,7H-pyrazolo[1,5-a]pyridine-3-carboxylate

  • MDL number MFCD26130364
  • Cas Number : 1779126-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1721909

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721909
Availability details for
Quantity
  • Availability
  • Add to Cart