ENAH2C55738B Enamine

5-(1-methyl-1H-pyrazol-4-yl)pyrazolidin-3-one

  • MDL number MFCD31667874
  • Cas Number : 2013869-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1721969

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721969
Availability details for
Quantity
  • Availability
  • Add to Cart