ENAH2C55738D Enamine

1-(oxan-4-yl)piperazin-2-one hydrochloride

  • MDL number MFCD22581540
  • Cas Number : 1284244-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009791

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009791
Availability details for
Quantity
  • Availability
  • Add to Cart