ENAH2C55738F Enamine

1-{8,8-dimethyl-1-oxaspiro[4.5]decan-2-yl}methanamine hydrochloride

  • MDL number MFCD31666896
  • Cas Number : 2230798-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009799

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009799
Availability details for
Quantity
  • Availability
  • Add to Cart