ENAH2C557390 Enamine

(2S)-4-hydroxy-2-(trimethylazaniumyl)butanoate

  • MDL number MFCD31667323
  • Cas Number : 97806-64-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009803

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009803
Availability details for
Quantity
  • Availability
  • Add to Cart