ENAH2C557391 Enamine

6,8-dichloro-4-hydroxyquinoline-3-carbonitrile

  • MDL number MFCD13181156
  • Cas Number : 61338-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009804

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009804
Availability details for
Quantity
  • Availability
  • Add to Cart