ENAH2C557393 Enamine

2-(prop-2-yn-1-yl)oxolane-2-carboxylic acid

  • MDL number MFCD29494842
  • Cas Number : 1936666-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009807

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009807
Availability details for
Quantity
  • Availability
  • Add to Cart