ENAH2C557396 Enamine

methyl 5-bromo-4-chloropyridine-2-carboxylate

  • MDL number MFCD18257817
  • Cas Number : 1256823-57-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1722041

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722041
Availability details for
Quantity
  • Availability
  • Add to Cart