ENAH2C557427 Enamine

[(1r,3r)-1-(aminomethyl)-3-(benzyloxy)cyclobutyl]methanol hydrochloride

  • MDL number MFCD31667326
  • Cas Number : 2230789-81-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009901

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009901
Availability details for
Quantity
  • Availability
  • Add to Cart