ENAH2C5574B5 Enamine

4-bromo-1H-pyrrole-3-carbaldehyde

  • MDL number MFCD20677048
  • Cas Number : 1378850-57-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009945

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009945
Availability details for
Quantity
  • Availability
  • Add to Cart