ENAH2C5574BE Enamine

4-[(1H-1,2,3-triazol-5-yl)methyl]piperidine-4-carboxylic acid dihydrochloride

  • MDL number MFCD31667332
  • Cas Number : 2230803-81-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009951

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009951
Availability details for
Quantity
  • Availability
  • Add to Cart