ENAH2C5574DC Enamine

1,3,2lambda6-benzodioxathiole-2,2-dione

  • MDL number MFCD00015885
  • Cas Number : 4074-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 33-36
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009882

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009882
Availability details for
Quantity
  • Availability
  • Add to Cart