ENAH2C557504 Enamine

2-(propan-2-yl)-1,3-oxazolidine

  • MDL number MFCD19216991
  • Cas Number : 27035-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009912

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009912
Availability details for
Quantity
  • Availability
  • Add to Cart