ENAH2C557555 Enamine

2-(3,3-difluoropiperidin-4-yl)acetonitrile

  • MDL number MFCD31666900
  • Cas Number : 1782516-55-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009916

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009916
Availability details for
Quantity
  • Availability
  • Add to Cart