ENAH2C5575B1 Enamine

2-(trifluoromethyl)-4,5,6,7-tetrahydro-1H-1,3-benzodiazole-5-carboxylic acid

  • MDL number MFCD24267800
  • Cas Number : 1472029-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009921

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009921
Availability details for
Quantity
  • Availability
  • Add to Cart