ENAH2C5576FE Enamine

tert-butyl 4-(hydroxymethyl)-4-phenylpiperidine-1-carboxylate

  • MDL number MFCD11044987
  • Cas Number : 158144-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-4353972

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-4353972
Availability details for
Quantity
  • Availability
  • Add to Cart