ENAH2C557729 Enamine

1-[5-(4-chlorophenyl)-1,2,4-oxadiazol-3-yl]methanamine hydrochloride

  • MDL number MFCD08733174
  • Cas Number : 1179370-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009834

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009834
Availability details for
Quantity
  • Availability
  • Add to Cart