ENAH2C55779E Enamine

3-[1-(aminomethyl)cyclopropyl]-1,2-oxazole-4-carboxylic acid hydrochloride

  • MDL number MFCD31666901
  • Cas Number : 2230802-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009934

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009934
Availability details for
Quantity
  • Availability
  • Add to Cart