ENAH2C5577A3 Enamine

3,6-dimethyl-1H-indole-2-carboxylic acid

  • MDL number MFCD16875904
  • Cas Number : 1260913-57-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009900

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009900
Availability details for
Quantity
  • Availability
  • Add to Cart