ENAH2C558170 Enamine

7-methoxy-1,2,3,4-tetrahydronaphthalen-2-amine hydrochloride

  • MDL number MFCD09026745
  • Cas Number : 3880-78-2
  • Molecular Weight : 213.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6481392

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6481392
Availability details for
Quantity
  • Availability
  • Add to Cart