ENAH2C558230 Enamine

tert-butyl N-[(1S)-1-(3-aminophenyl)ethyl]carbamate

  • MDL number MFCD31668283
  • Cas Number : 1610767-01-5
  • Molecular Weight : 236.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6482458

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6482458
Availability details for
Quantity
  • Availability
  • Add to Cart