ENAH3035CA86 Enamine

1H,2H,3H,4H-pyrido[3,4-d]pyrimidine-2,4-dione

  • MDL number MFCD07438024
  • Cas Number : 21038-67-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-106860

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106860
Availability details for
Quantity
  • Availability
  • Add to Cart