ENAH3035CA8C Enamine

ethyl 4-chloro-2-(propan-2-yl)pyrimidine-5-carboxylate

  • MDL number MFCD11047086
  • Cas Number : 72419-33-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-107579

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107579
Availability details for
Quantity
  • Availability
  • Add to Cart