ENAH3035CA92 Enamine

8-methoxy-1-methyl-2,3,4,5-tetrahydro-1H-3-benzazepin-2-one

  • MDL number MFCD24447799
  • Cas Number : 849663-11-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-108454

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108454
Availability details for
Quantity
  • Availability
  • Add to Cart