ENAH3035CA9A Enamine

(1,4-dimethyl-1H-imidazol-2-yl)methanol

  • MDL number MFCD11110199
  • Cas Number : 59608-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-110068

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110068
Availability details for
Quantity
  • Availability
  • Add to Cart