ENAH3035CAA9 Enamine

3-methoxy-5-methyl-1,2-oxazole-4-carboxylic acid

  • MDL number MFCD18820497
  • Cas Number : 16877-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-112833

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112833
Availability details for
Quantity
  • Availability
  • Add to Cart