ENAH3035CAB8 Enamine

2-{[(tert-butoxy)carbonyl]amino}-2-(1-methyl-1H-pyrazol-4-yl)acetic acid

  • MDL number MFCD19401000
  • Cas Number : 1394970-92-3
  • Molecular Weight : 255.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-114990

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114990
Availability details for
Quantity
  • Availability
  • Add to Cart