ENAH3035CAC9 Enamine

5-(chloromethyl)-3-phenyl-1H-pyrazole hydrochloride

  • MDL number MFCD29045842
  • Cas Number : 493038-58-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 171-172
Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-117969

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117969
Availability details for
Quantity
  • Availability
  • Add to Cart