ENAH3035CB09 Enamine

2-methylbutane-2-sulfonyl chloride

  • MDL number MFCD19200188
  • Cas Number : 16489-14-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-126042

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126042
Availability details for
Quantity
  • Availability
  • Add to Cart