ENAH3035CB0F Enamine

[4-(2-methylpropyl)phenyl]methanol

  • MDL number MFCD11111048
  • Cas Number : 110319-85-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-127716

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127716
Availability details for
Quantity
  • Availability
  • Add to Cart