ENAH3035CB11 Enamine

1-(4-{[(tert-butoxy)carbonyl]amino}phenyl)cyclopropane-1-carboxylic acid

  • MDL number MFCD19693623
  • Cas Number : 1314778-71-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-127948

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127948
Availability details for
Quantity
  • Availability
  • Add to Cart