ENAH3035CB12 Enamine

1-[4-(chlorosulfonyl)phenyl]cyclopropane-1-carboxylic acid

  • MDL number MFCD28536974
  • Cas Number : 1780234-78-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-127949

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127949
Availability details for
Quantity
  • Availability
  • Add to Cart