ENAH3035CB1F Enamine

5-chloro-3-methyl-1-(propan-2-yl)-1H-pyrazole

  • MDL number MFCD21667695
  • Cas Number : 1480458-98-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-132328

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132328
Availability details for
Quantity
  • Availability
  • Add to Cart