ENAH3035CB30 Enamine

ethyl 1-(2-cyanoethyl)-2-oxocyclohexane-1-carboxylate

  • MDL number MFCD19302705
  • Cas Number : 42894-09-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-136688

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136688
Availability details for
Quantity
  • Availability
  • Add to Cart