ENAH3035CB35 Enamine

1-(1H-1,2,4-triazol-3-yl)ethan-1-one

  • MDL number MFCD18834711
  • Cas Number : 153334-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158-163
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-137277

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137277
Availability details for
Quantity
  • Availability
  • Add to Cart