ENAH3035CB36 Enamine

1-(4H-1,2,4-triazol-3-yl)propan-1-one

  • MDL number MFCD21194679
  • Cas Number : 1480235-50-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-137295

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137295
Availability details for
Quantity
  • Availability
  • Add to Cart