ENAH3035CB54 Enamine

methyl imidazo[1,2-a]pyrimidine-6-carboxylate

  • MDL number MFCD11977222
  • Cas Number : 1083196-24-0
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-141350

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141350
Availability details for
Quantity
  • Availability
  • Add to Cart